Would you like to log in again?
VWR® Temperature Monitoring Device
Supplier: VWR
VWR®
CP-TMD-18
76520-150EA 155.5 USD InStock
76520-150
VWR® Temperature Monitoring Device
Refrigerators
This temperature monitoring device comes with a buffered probe in product simulated solution. It includes an °F/°C switchable readout, audible and visual alarms, and a 3 year certificate of calibration.
  • Material: Assorted materials
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty