Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diamond® SureFlow™ Bottle Top Dispensers, Globe Scientific

Supplier: Globe Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

GBTD-R-2.5 GBTD-25 GBTD-10 GBTD-100 GBTD-30 GBTD-R-25 GBTD-R-05 GBTD-2.5 GBTD-05 GBTD-R-10 GBTD-R-50 GBTD-R-30 GBTD-60 GBTD-R-100 GBTD-R-60 GBTD-50
76520-464EA 335.21 USD InStock
76520-464 76520-474 76520-466 76520-476 76520-454 76520-468 76520-456 76520-478 76520-458 76520-480 76520-460 76520-482 76520-470 76520-462 76520-484 76520-472
Diamond® SureFlow™ Bottle Top Dispensers, Globe Scientific
Bottle Top Dispensers
Diamond® SureFlow™ Bottle Top Dispensers have been designed with several features to make the dispensing of valuable laboratory reagents easier than ever.

  • Two-year warranty
  • Fully autoclavable
  • Single tool recalibration
  • Locking feature to prevent spills
  • 360° rotation to dispense in multiple vessels
  • Lower pressure for larger volumes helps reduce splashing
  • Telescopic fill tube for bottles ranging in height from 170 to 300 mm

With a choice of standard or recirculating models, bottle top dispensers include a unique piston design which allows cleaning and servicing without changing the calibration, and a springless valve which requires less maintenance, offering higher reliability and added chemical resistance. Dual graduations, volume lock, and four additional bottle thread adapters are all standard. Recirculating models help eliminate the loss of valuable reagents by priming without loss.

Order Now
 

ORDER

ACCESSORIES

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty