Would you like to log in again?
Tyvek® 1073B Autoclave Bags with Steam Indicator, Self-Seal, Keystone Cleanroom Products
Supplier: Keystone Cleanroom Products
COVMGTVK3036SS
76520-574CS 761.09 USD InStock
76520-574
Tyvek® 1073B Autoclave Bags with Steam Indicator, Self-Seal, Keystone Cleanroom Products
Bags Sterilization Pouches
Self-Sealing 1073B Tyvek® are ideal for steam sterilization of pharmaceutical components such as vial and syringe stoppers and caps as well as heavy equipment parts with sharp edges. No need to use or validate a heat sealing machine.
  • Manufactured and packaged in a controlled cleanroom environment
  • Convenient Self-sealing closure system
  • Autoclavable at 121 to 125 °C
  • Tear and puncture resistant, Hydrophobic and lint-free
  • 100% Medical Grade 1073B Tyvek® permits fast steam penetration
  • Excellent impact resistance
  • Consistently strong factory seals, Exceptional microbial barrier
100% Medical Grade self-sealing 1073B Tyvek® Autoclave bags are ideal for steam sterilization of pharmaceutical components such as vial and syringe stoppers and caps.
With consistently strong factory seals, these are a good choice for heavy, or pharmaceutical change parts with sharp corners. All Tyvek construction allows for a greater number of bags per sterilization load; increasing efficiencies and cost reduction.
Self-sealing closure reduces prep time and eliminates the need for and the validation of a heat sealing machine.
Certifications: Certificates of Conformance sent with each shipment.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty