Would you like to log in again?
Evergreen One Slide Mailers, Caplugs Evergreen
Supplier: Caplugs Evergreen
240-3071-030 240-3071-B80
76520-584BG 18.66 USD InStock
76520-584 76520-582
Evergreen One Slide Mailers, Caplugs Evergreen
Slide Mailers
Caplugs Evergreen one-slide mailers feature a sturdy living hinge design with friction-fit and post-type locks.
  • Top-panel window allows slides to be viewed without opening
  • Easy-to-use finger/thumb tabs eliminate prying and/or cutting tape
  • Available in multiple colors
A large matte-finished area is molded-in to provide ample room for identification. The slim shape makes it ideal as a component of transport and mailing kits.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty