Would you like to log in again?
24-Well Plates, Caplugs
Supplier: Caplugs
290-8324-03F 222-8044-01F 290-8124-01F 333-8024-01F
76520-642CT 271.82 USD InStock
76520-642 76520-644 76520-634 76520-636
24-Well Plates, Caplugs
Microplates Deep Well Microplates
24-well plates are gamma-irradiated for sterility and available in both sterile and treated versions.
  • Distinct alpha-numeric well designations
  • Well volume: 3.5 ml
  • Available with or without lid
Available in individually packed units.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty