Would you like to log in again?
BT WIRELESS LOGGER: -200TO+50C
Supplier: Thermo Fisher Scientific
Wireless datalogging module, with external temperature PT100 probe, range: -200 deg C to +50 deg C, 1m in length, Calibrated: -80 deg C. Package consists of: Module, External Sensor, Lithium Battery, and 1 magnetic mount.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BT WIRELESS LOGGER: -200TO+50C 76520-810 SVPWSENTCS002 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty