Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Excel EX444 Crucibles

Supplier: Morgan Advanced Materials
Excel
69440000000444
76520-932EA 705.61 USD InStock
76520-932
Excel EX444 Crucibles
Crucibles
Straight shape silicon carbide crucibles for induction furnaces.

Brass capacity: 90 kg

Brimful capacity water: 15 L

Pyrometer pocket and hole in wall configurations are available to facilitate measurement of metal temperature.

Recommended for non-ferrous alloys except thosecontaining more than 30% of Nickel, Chromium or Iron.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty