Would you like to log in again?
Centrifuge Tubes, Caplugs
Supplier: Caplugs
220-3530-030 220-3928-080
76520-950CT 229.68 USD InStock
76520-950 76520-948
Centrifuge Tubes, Caplugs
Tubes Centrifuge Tubes
Centrifuge tubes are ideal for general purpose laboratory work. These tubes are especially useful as a leak-proof primary container in shipping infectious specimens.
  • Made of polypropylene
  • Nearly transparent
  • Withstand up to 10000×g
  • With conical bottom or freestanding octagonal base that lock into the special tray for one-hand operation
Appropriate for use with solvents, alcohols, chlorinated and aromatic hydrocarbons (except xylene) and ketones.
Ordering information: Screw caps can be ordered separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty