Would you like to log in again?
Evergreen 24-Well Deep Well Plates, Caplugs Evergreen
Supplier: Caplugs Evergreen
222-8685-08C 290-8573-08C
76521-422CT 125.31 USD InStock
76521-422 76521-420
Evergreen 24-Well Deep Well Plates, Caplugs Evergreen
Microplates Deep Well Microplates
Caplugs evergreen 24-well deep-well plates have the same footprint as the regular microplates and are available in sterile and non sterile versions.
  • Suitable for dilution and storage
  • Distinct alpha-numeric well designations
  • Well volume: 10 ml
  • Flat bottom with chamfered edges
  • Polypropylene lids are available
  • EZ Seal™ film compatible
48 deep-well and 96 deep-well units are also available.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty