Would you like to log in again?
RESPIRATOR PARTICULAT 9205 N95 BLK CS440
Supplier: 3M OCCUPATIONAL HEALTH
Particulate Respirator 9205+, N95, Bulk, Non-Oil, White, Respirator Size: Standard, Respirator Style: Flat Fold, Strap Attachment Type: Stapled, Designed to direct exhaled air away from the nose panel, helping reduce eyewear fogging
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
RESPIRATOR PARTICULAT 9205 N95 BLK CS440 76521-452 7100232940 Case of 440 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty