Would you like to log in again?
RESPIRATOR KIMTECH N95 POUCH SZ SM CS300
Supplier: KIMBERLY CLARK CORP
respirator, NIOSH-certified N95 particulate filter, provides a minimum 95% filtration efficiency of 0.3 micron particles (though not oil resistant), has not been tested vs COVID-19 and can’t be recommended for efficacy or use, NOT for medical use. Size Small
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
RESPIRATOR KIMTECH N95 POUCH SZ SM CS300 76521-766 54066 Case of 300 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty