Would you like to log in again?
TouchNTuff® 83-500 Sterile Polyisoprene Gloves, Ansell
Supplier: Ansell
TouchNTuff®
83500090 83500055 83500065 83500085 83500070 83500090 83500085 83500065 83500060 83500080 83500075 83500070 83500075 83500055 83500080 83500060
76521-780CS 467.93 USD InStock
76521-780 CA76521-440 CA76521-442 CA76521-444 CA76521-434 76521-776 76521-774 76521-772 76521-782 76521-770 76521-768 76521-778 CA76521-446 CA76521-436 CA76521-448 CA76521-438
TouchNTuff® 83-500 Sterile Polyisoprene Gloves, Ansell
Gloves
TouchNTuff® 83-500 is an ultra-soft, sterile polyisoprene glove suitable for Class 100 (ISO 5) cleanroom environments. It provides a comfortable, 'second skin' feel for longer wear time and reduced hand fatigue.
  • Superior comfort with 'second skin' feel
  • Beaded cuffs with SUREFIT™ Technology
  • Latex-free polyisoprene performance
  • Prevents Type I allergies
The latex-free polyisoprene material offers the performance of natural rubber latex without the risk of latex sensitization. Ideal for double donning. The gloves are best suited for use in life sciences and chemical industries.
Certifications: AS/NZS 4179, ASTM D3577, ASTM D7160, CE 0493, EN 420:2003 + A1:2009, EN 556-1:2001, EN ISO 374-1:2016, EN ISO 374-5:2016, EN 374:2003, EN 421:2010. Please consult the IFU. ISO 10282, ISO 11193. Category III. Compatible with Class 100 (ISO 5)/Grade A cleanroom environments.
Packaging: Gloves are packaged 1 pair per inner polybag; 10 pairs per outer polybag; 5 polybags per master polybags; 4 master polybags per carton; 200 pairs per carton.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty