Would you like to log in again?
Abdos® Cryo Vials, Polyproplyene, Gamma Sterilized, Foxx Life Sciences
Supplier: Foxx Life Sciences
P60109 P60112 P60108 P60118 P60115 P60116 P60117
76521-938CS 523.55 USD InStock
76521-938 76521-936 76521-934 76521-956 76521-954 76521-952 76521-950
Abdos® Cryo Vials, Polyproplyene, Gamma Sterilized, Foxx Life Sciences
Vials
These cryo vials are manufactured using high purity virgin polypropylene (PP), conforming to US FDA 21 CFR standards, which is free of rubbers and heavy metals. Designed for long-term cryogenic storage of biological samples, all cell types, enzymes, or reagents.
  • Material: Polypropylene
  • Temperature range: –196...+126 °C
  • Sterilization by radiation: Gamma
  • Material transparency: Transparent
  • Autoclavable: Yes
  • Shelf life: 3 years
The leak-proof cap design and strong wall thickness make it an ideal product for transporting samples and specimens. These cryo vials are designed for mechanical freezing and its screw cap reduces the risk of contamination during handling.
The large white frosted area on the surface allows for ease of labeling/writing. Graduation in silk-screen printing makes identification easy.
Certifications: Complies with: ISO 11137. Complies with: ISO Standard: 9001, 13485, 14001. Batch certificate included.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty