Would you like to log in again?
UVP SOLO ELITE 6.3MP COLOR 115V
Supplier: Analytik Jena US
Uvp Solo Elite, 6.3Mp Color, EtBr emission filter pre-mounted on the Elite Mount, UVP Elite UV Transilluminator (302 nm), Resoultion: 3072 x 2048 pixel, from fluorescent DNA gels, fluorescent protein gels, and blue light gels, to colorimetry, Voltage: 115V
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
UVP SOLO ELITE 6.3MP COLOR 115V 76522-044 849-97-0944-03 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty