Would you like to log in again?
Globe Glass™ Graduated Cylinders, To Deliver, Globe Scientific
Supplier: Globe Scientific
8300250 8331000 8302000 8330010 8330250 8300050 8300100 8330025 8301000 8330100 8300500 8330005 8330500 8300010 8300005 8330050 8332000 8300025
76523-342BX 105.4 USD InStock
76523-342 76523-320 76523-330 76523-340 76523-350 76523-324 76523-346 76523-334 76523-344 76523-322 76523-332 76523-328 76523-338 76523-316 76523-326 76523-348 76523-336 76523-318
Globe Glass™ Graduated Cylinders, To Deliver, Globe Scientific
Cylinders Measuring Cylinders
Globe Glass™ graduated cylinders are calibrated to ASTM E1272 standards for precise measurement of volumes of liquids. A solid hexagonal base provides stability, and an included polyethylene (PE) bumper guard helps to protect against accidental breakage. These graduated cylinders are calibrated TD, 'To Deliver', and are available in sizes ranging from 5 to 2000 ml, with Class A or Class B calibration tolerance options. Class A graduated cylinders include a batch calibration certificate.
  • Calibrated TD; 'To Deliver'
  • Class A calibration tolerance for highest accuracy and the most critical laboratory work
  • Class B calibration tolerance for general laboratory work
  • Pour spout for easy liquid transfer
  • Beaded rim for strength and durability
  • High resistance to chemicals
  • Dual scale metric graduations; durable white enamel printing
Globe Glass™ lab glassware is made exclusively from ASTM E438, Type I, Class A, 3.3 borosilicate glass for excellent resistance to thermal shock and high chemical resistance to most acids, alkalis, and solvents. Globe Glass™ lab glassware meets applicable ASTM standards for volume, shape, dimensions, graduation markings, and calibration. These ASTM standards are clearly marked for easy confirmation that each piece of laboratory glassware will meet the needs of laboratory professionals. Computer controlled, automated production lines ensure that this laboratory glassware has consistent wall thickness, flat bottoms free of striations, and precise forming of rims and spouts.
Permanent white enamel markings are ASTM compliant and easy to read. Dual metric scale graduations are included on most items for easy reference while filling or dispensing. A large, convenient marking area is provided for coding or identification. Globe Glass™ laboratory glassware is manufactured under stringent quality controls. Manufacturing facilities are ISO 9001 certified and in-house calibration is performed in an ISO 17025 certified laboratory.
Certifications: Cylinders meet ASTM E1272 Style 1 specification.
Packaging: GlassGuard™ Packaging is engineered to meet the hazards and rigors of transportation, protecting Globe Glass laboratory glassware during shipment. Robust packaging designs begin at the pack level and extend to the full case packaging. Heavy-duty, multi-layered corrugated container systems meet rigid quality standards and testing protocols to provide exceptional protection from damage, even in the small parcel shipping environment.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty