Would you like to log in again?
PEN CIRCUITWORKS LEAD FREE DISPENSING 9G
Supplier: ITW-CHEMTRONICS,INC
Pen, Circuitworks, Lead Free, Dispensing, Flux, quickly applies a noncorrosive, halide-free, No Clean flux, Size: 9g
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
PEN CIRCUITWORKS LEAD FREE DISPENSING 9G 76523-352 CW8400 Case of 12 Retrieving
Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty