Would you like to log in again?
AseptiQuik® Sterile Connector Cuffs
Supplier: CPC (Colder Products Company)
CPC® AseptiQuik® S CPC® AseptiQuik® G CPC® AseptiQuik® L
AQSCUFF AQLCUFF AQGCUFF
76523-386EA 58.15 USD InStock
76523-386 76523-384 76523-388
AseptiQuik® Sterile Connector Cuffs
Fittings and Connectors
AseptiQuik® Connector Cuffs provide additional support for more robust applications, so users can continue to transfer media easily with these quick and easy to use AseptiQuik® sterile connectors.
  • Genderless design eases single-use system specifications with one-part number for both halves
  • Flip-Click-Pull: intuitive three-step connection process reduces risk of operator error
  • Audible confirmation of connection with no additional hardware required
  • BPA-free
Use the AseptiQuik® cuffs to support use cases which have excessive sideload, pump, tension, vibration, or reduction in flow.
AseptiQuik® PPSU enables genderless connection for a greater range of chemical applications, offering versatile connections across downstream processes with a pH range from 2 to 12.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty