VHB™ Tapes are fast and easy-to-use, with a permanent bonding method to provide high strength and long-term durability.
- White, 25 mil (0.6 mm), general-purpose adhesive and firm acrylic foam core
- Permanent bonding provides high strength and long-term durability
- Can replace mechanical fasteners (rivets, welds, screws) or liquid adhesives
- Maximum temperature: 300 °F (149 °C)
- Adhesive strength: 20 lbs./inch
VHB™ Tape 4930 is a firm, double-sided, pressure-sensitive, closed-cell acrylic foam tape. High-strength and durable, with a clean appearance.