Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VHB™ 3M™ 4930 Tapes, TapeCase

Supplier: TAPECASE LTD. TE

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11
76523-580PK 72.11 USD InStock
76523-580 76523-584 76523-462 76523-582 76523-460 76523-588 76523-500 76523-620 76523-466 76523-586 76523-464 76523-614 76523-458 76523-612 76523-578 76523-618 76523-616 76523-470 76523-590 76523-474 76523-594 76523-472 76523-592 76523-478 76523-632 76523-598 76523-510 76523-476 76523-630 76523-596 76523-504 76523-624 76523-502 76523-468 76523-622 76523-508 76523-628 76523-506 76523-626 76523-480 76523-484 76523-482 76523-522 76523-642 76523-488 76523-520 76523-640 76523-486 76523-636 76523-514 76523-634 76523-512 76523-518 76523-638 76523-516 76523-492 76523-490 76523-650 76523-496 76523-494 76523-654 76523-412 76523-532 76523-652 76523-498 76523-530 76523-526 76523-646 76523-524 76523-644 76523-528 76523-648 76523-540 76523-660 76523-544 76523-422 76523-664 76523-542 76523-420 76523-662 76523-658 76523-416 76523-536 76523-414 76523-656 76523-534 76523-418 76523-538 76523-672 76523-430 76523-550 76523-670 76523-434 76523-554 76523-432 76523-552 76523-548 76523-426 76523-668 76523-546 76523-424 76523-666 76523-428 76523-562 76523-440 76523-560 76523-566 76523-444 76523-564 76523-442 76523-438 76523-558 76523-436 76523-556 76523-452 76523-572 76523-450 76523-570 76523-610 76523-456 76523-576 76523-454 76523-574 76523-448 76523-602 76523-568 76523-446 76523-600 76523-606 76523-604 76523-608
VHB™ 3M™ 4930 Tapes, TapeCase
Tapes
VHB™ Tapes are fast and easy-to-use, with a permanent bonding method to provide high strength and long-term durability.

  • White, 25 mil (0.6 mm), general-purpose adhesive and firm acrylic foam core
  • Permanent bonding provides high strength and long-term durability
  • Can replace mechanical fasteners (rivets, welds, screws) or liquid adhesives
  • Maximum temperature: 300 °F (149 °C)
  • Adhesive strength: 20 lbs./inch

VHB™ Tape 4930 is a firm, double-sided, pressure-sensitive, closed-cell acrylic foam tape. High-strength and durable, with a clean appearance.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty