VHB™ Tapes are fast and easy-to-use, with a permanent bonding method to provide high strength and long-term durability.
- White, 1.1 mm (45 mil), general-purpose adhesive and firm acrylic foam core
- Permanent bonding method provides high strength and long-term durability
- Can replace mechanical fasteners (rivets, welds, screws) or liquid adhesives
- Maximum temperature: 149 °C (300 °F)
- Adhesive strength: 25 lbs./inch
VHB™ Tape 4950 replaces screws, rivets, and studs to permanently adhere one substrate to another while spreading the stress load across the entire length of the joint. Once materials are joined with this tape, a virtually indestructible weld is created. It combines the most conformable acrylic foam with modified acrylic adhesive.