Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VHB™ 3M™ 4950 Tapes, Tapecase

Supplier: TAPECASE LTD. TE

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11
76523-980PK 10.75 USD InStock
76523-980 76523-984 76523-742 76523-862 76523-982 76523-740 76523-860 76523-856 76523-976 76523-734 76523-854 76523-974 76523-732 76523-738 76523-858 76523-736 76523-978 76523-870 76523-990 76523-874 76523-752 76523-994 76523-872 76523-750 76523-992 76523-746 76523-900 76523-988 76523-866 76523-986 76523-744 76523-864 76523-904 76523-902 76523-748 76523-868 76523-908 76523-906 76523-760 76523-880 76523-764 76523-884 76523-762 76523-882 76523-878 76523-756 76523-998 76523-910 76523-876 76523-996 76523-754 76523-914 76523-758 76523-912 76523-918 76523-916 76523-892 76523-770 76523-890 76523-896 76523-774 76523-894 76523-772 76523-768 76523-922 76523-888 76523-800 76523-766 76523-920 76523-886 76523-926 76523-804 76523-924 76523-802 76523-808 76523-928 76523-806 76523-782 76523-780 76523-786 76523-940 76523-784 76523-812 76523-778 76523-932 76523-898 76523-810 76523-776 76523-930 76523-816 76523-936 76523-814 76523-934 76523-818 76523-938 76523-792 76523-790 76523-830 76523-796 76523-950 76523-794 76523-944 76523-702 76523-822 76523-788 76523-700 76523-942 76523-820 76523-948 76523-706 76523-826 76523-946 76523-704 76523-824 76523-708 76523-828 76524-006 76524-008 76523-682 76523-962 76523-720 76523-840 76523-686 76523-960 76523-684 76523-834 76523-712 76523-954 76523-832 76523-952 76523-798 76523-710 76523-838 76523-958 76523-716 76523-836 76523-956 76523-714 76523-718 76524-010 76523-690 76523-694 76523-692 76523-698 76523-852 76523-972 76523-730 76523-850 76523-696 76523-970 76523-724 76523-966 76523-844 76523-722 76523-964 76523-842 76523-688 76523-728 76523-848 76523-726 76523-968 76523-846 76524-000 76524-002 76524-004
VHB™ 3M™ 4950 Tapes, Tapecase
Tapes
VHB™ Tapes are fast and easy-to-use, with a permanent bonding method to provide high strength and long-term durability.

  • White, 1.1 mm (45 mil), general-purpose adhesive and firm acrylic foam core
  • Permanent bonding method provides high strength and long-term durability
  • Can replace mechanical fasteners (rivets, welds, screws) or liquid adhesives
  • Maximum temperature: 149 °C (300 °F)
  • Adhesive strength: 25 lbs./inch

VHB™ Tape 4950 replaces screws, rivets, and studs to permanently adhere one substrate to another while spreading the stress load across the entire length of the joint. Once materials are joined with this tape, a virtually indestructible weld is created. It combines the most conformable acrylic foam with modified acrylic adhesive.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty