Would you like to log in again?
VHB™ 3M™ 4955 Tapes, TapeCase
Supplier: TAPECASE LTD. TE
8.88519E+11 8.88519E+11 8.88519E+11
76524-016PK 155.31 USD InStock
76524-016 76524-012 76524-014
VHB™ 3M™ 4955 Tapes, TapeCase
Tapes
VHB™ Tapes are fast and easy-to-use, with a permanent bonding method to provide high strength and long-term durability.
  • White, 2.0 mm (80 mil), general-purpose adhesive and firm acrylic foam core
  • Permanent bonding method provides high strength and long-term durability
  • Can replace mechanical fasteners (rivets, welds, screws) or liquid adhesives
  • Maximum temperature: 204 °C (400 °F)
  • Adhesive strength: 20 lbs./inch
This white, 2.0 mm (0.080"), tape features a general-purpose acrylic adhesive with a firm foam core. It can replace rivets, welds, and screws. It also offers design flexibility with its viscoelasticity and powerful ability to bond to a variety of surfaces.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty