VHB™ tapes are fast and easy-to-use, with a permanent bonding method to provide high strength and long-term durability.
- Black, 45 mil thick, (1.1 mm), modified acrylic adhesive and very conformable acrylic foam core
- Permanent bonding method provides high strength and long-term durability
- Can replace mechanical fasteners (rivets, welding, screws) or liquid adhesives
- Maximum temperature: 300 °F (149 °C)
- Adhesive strength: 22 lbs./inch
VHB™ tape 5952 is very conformable double-sided foam tape. This tape is designed for bonding with medium low surface energy paints, plastics, and powder coated paints. With an extra conformable foam construction, this product offers the maximum performance when adapting to irregular or mismatched bonding surfaces.