Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VHB™ 3M™ GPH-060GF Tapes, TapeCase

Supplier: TAPECASE LTD. TE

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11
76524-544PK 124.52 USD InStock
76524-544 76524-566 76524-522 76524-524 76524-546 76524-568 76524-526 76524-548 76524-528 76524-506 76524-508 76524-570 76524-572 76524-550 76524-574 76524-530 76524-552 76524-510 76524-576 76524-532 76524-554 76524-534 76524-556 76524-512 76524-536 76524-514 76524-558 76524-538 76524-516 76524-518 76524-560 76524-540 76524-562 76524-520 76524-542 76524-564
VHB™ 3M™ GPH-060GF Tapes, TapeCase
Tapes
VHB™ tapes are fast and easy-to-use, with a permanent bonding method to provide high strength and long-term durability.

  • Gray, 25 mil, 0.025" (0.6 mm) thick, acrylic adhesive with a conformable, acrylic foam core
  • Permanent bonding method provides high strength and long-term durability
  • Can replace mechanical fasteners (rivets, welding, screws) or liquid adhesives in appropriate applications
  • Maximum temperature: 450 °F (232 °C)
  • Adhesive strength: 14 lbs./inch

VHB™ tape GPH-060GF is double-sided acrylic foam tape with a red polyethylene film liner. The conformable foam provides good contact between substrates even when they are slightly mismatched.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty