Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VHB™ 3M™ GPH-110GF Tapes, TapeCase

Supplier: TAPECASE LTD. TE

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11
76524-600RL 20.79 USD InStock
76524-600 76524-622 76524-588 76524-602 76524-624 76524-626 76524-604 76524-606 76524-608 76524-590 76524-592 76524-594 76524-596 76524-598 76524-578 76524-610 76524-612 76524-614 76524-616 76524-618 76524-580 76524-582 76524-584 76524-620 76524-586
VHB™ 3M™ GPH-110GF Tapes, TapeCase
Tapes
VHB™ Tapes are fast and easy-to-use, with a permanent bonding method to provide high strength and long-term durability.

  • Gray, 1.1 mm (45 mil), acrylic adhesive with a conformable, acrylic foam core
  • Permanent bonding method provides high strength and long-term durability
  • Can replace mechanical fasteners (rivets, welding, screws) or liquid adhesives in appropriate applications
  • Maximum temperature: 232 °C (450 °F)
  • Adhesive strength: 21 lbs./inch

VHB™ Tape GPH-110GF is a 1.1 mm (0.045") thick, gray double-sided acrylic foam tape with a red polyethylene film liner. The conformable foam provides good contact between substrates even when they are slightly mismatched.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty