VHB™ tapes are fast and easy-to-use, with a permanent bonding method to provide high strength and long-term durability.
- Gray, 62 mil, 0.062" (1.6 mm) thick, acrylic adhesive with a conformable, acrylic foam core
- Permanent bonding method provides high strength and long-term durability
- Can replace mechanical fasteners (rivets, welding, screws) or liquid adhesives in appropriate applications
- Maximum temperature: 450 °F (232 °C)
- Adhesive strength: 19 lbs./inch
VHB™ tape GPH-160GF is double-sided acrylic foam tape with a red polyethylene film liner. The conformable foam provides good contact between substrates even when they are slightly mismatched.