Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VHB™ 3M™ GPH-160GF Tapes, TapeCase

Supplier: TAPECASE LTD. TE

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11
76524-688RL 344.77 USD InStock
76524-688 76524-666 76524-644 76524-720 76524-668 76524-646 76524-700 76524-648 76524-702 76524-628 76524-704 76524-706 76524-708 76524-690 76524-670 76524-692 76524-672 76524-650 76524-694 76524-630 76524-674 76524-696 76524-652 76524-710 76524-632 76524-654 76524-698 76524-676 76524-712 76524-656 76524-634 76524-678 76524-714 76524-636 76524-658 76524-716 76524-638 76524-718 76524-680 76524-660 76524-682 76524-640 76524-684 76524-662 76524-642 76524-664 76524-686
VHB™ 3M™ GPH-160GF Tapes, TapeCase
Tapes
VHB™ tapes are fast and easy-to-use, with a permanent bonding method to provide high strength and long-term durability.

  • Gray, 62 mil, 0.062" (1.6 mm) thick, acrylic adhesive with a conformable, acrylic foam core
  • Permanent bonding method provides high strength and long-term durability
  • Can replace mechanical fasteners (rivets, welding, screws) or liquid adhesives in appropriate applications
  • Maximum temperature: 450 °F (232 °C)
  • Adhesive strength: 19 lbs./inch

VHB™ tape GPH-160GF is double-sided acrylic foam tape with a red polyethylene film liner. The conformable foam provides good contact between substrates even when they are slightly mismatched.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty