Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VHB™ 3M™ LSE-110WF Tapes, TapeCase

Supplier: TAPECASE LTD. TE

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11
76524-742RL 28.47 USD InStock
76524-742 76524-744 76524-746 76524-738 76524-748 76524-750 76524-752 76524-740
VHB™ 3M™ LSE-110WF Tapes, TapeCase
Tapes
VHB™ Tapes are fast and easy-to-use, with a permanent bonding method to provide high strength and long-term durability.

  • White, 45 mil (1.1 mm) thick, modified acrylic adhesive and conformable acrylic foam core
  • Permanent bonding method provides high strength and long-term durability indoors and outdoors
  • Maximum temperature: 302 °F (150 °C)
  • Adhesive strength: 44 N/cm

VHB™ Tape LSE-110WF has modified acrylic adhesive on both sides of a conformable foam core. It can replace rivets, welds, and screws as well as other forms of mechanical fasteners or liquid adhesives.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty