Would you like to log in again?
Vikan® Hygienic Wall Bracket, Single Hook Module, Remco
Supplier: Remco
Vikan®
101266 10126 10121 10128 10125 101277 101288 10122 10127 10129 10123 10124
76528-638EA 12.35 USD InStock
76528-638 76528-628 76528-636 76528-626 76528-630 76528-642 76528-640 76528-634 76528-624 76528-646 76528-632 76528-644
Vikan® Hygienic Wall Bracket, Single Hook Module, Remco
Wall Hooks
Single hook module is designed for storing cleaning tools that have a hanging hole.
  • Can hold products weighing up to 6.61 lbs.
  • Polypropylene/polyamide construction
The hook system is easy to disassemble for cleaning or replacement.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty