Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Vikan® 16" UST Push Broom Head, Soft Bristles, Remco

Supplier: Remco
Vikan®
31717 31718 31716 31714 31711 31713 31712 31715
76528-836EA 59.23 USD InStock
76528-836 76528-826 76528-834 76528-824 76528-828 76528-832 76528-822 76528-830
Vikan® 16" UST Push Broom Head, Soft Bristles, Remco
Brooms
The 16" Ultra Safe Technology (UST) soft floor broom is effective for sweeping dry particles in high risk areas.

  • Polypropylene block, polyester bristles
  • Euro (hygienic) thread type

The angled arrangement of the Bristle Security Units ensures efficient and effective cleaning, while the lack of resin, glue, or staples means bristle units are easy to clean, FDA-compliant, and more secure.

Broom head dimensions: 15.25L×1.875W×4.7"H

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty