Would you like to log in again?
Vikan® UST Short Handle Scrubbing Brushes, Stiff Bristles, Remco
Supplier: Remco
Vikan®
41793 41798 41792 41797 41795 41791 41796 41794
76528-924EA 27.27 USD InStock
76528-924 76528-914 76528-922 76528-928 76528-918 76528-926 76528-916 76528-920
Vikan® UST Short Handle Scrubbing Brushes, Stiff Bristles, Remco
Brushes
This UST short handled hand brush with angled Bristle Security Units is effective when cleaning conveyor belts, production lines, machinery, and food preparation surfaces in high risk areas.
  • Polypropylene block, polyester bristles
Angled and circular arrangement of Bristle Security Units ensures effcient cleaning.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty