Would you like to log in again?
Vikan® Ultra-Slim Wand Brushes, Remco
Supplier: Remco
Vikan®
41975 41973 41976 41972 41974
76528-958EA 17.41 USD InStock
76528-958 76528-966 76528-960 76528-964 76528-962
Vikan® Ultra-Slim Wand Brushes, Remco
Brushes
Clean where other brushes cannot reach with this ultra-slim (15 mm wide) brush designed with a scraping edge for tackling stubborn debris.
  • Color-coded to support HACCP principles
  • Regenerated polypropylene block, polyester bristles
  • Food contact compliant
This brush features an easy-grip slim handle and a specially designed head with extra-stiff first bristles that help clean all the way to the end of narrow spaces. The brush block is made using food-contact compliant polypropylene offcuts from other Vikan products.
Overall dimensions: 23.62L×0.59W×2.36"H
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty