Would you like to log in again?
Vikan® High-Temp Hand Brush, Remco
Supplier: Remco
Vikan®
42885
76528-978EA 32.56 USD InStock
76528-978
Vikan® High-Temp Hand Brush, Remco
Brushes
Originally designed for removing debris from heat sealers and other packing equipment, this brush is also ideal for cleaning hot grills and frying pans.
  • Clean safely without contacting hot surfaces
  • Polypropylene block, polyester bristles
Ergonomically friendly design with a long handle for safe cleaning.
Overall dimensions: 11.42L×1.18W×2.56"H
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty