Would you like to log in again?
INCUBATOR 250 CR CO2 STACK ADAPTER PLATE
Supplier: Thermo Fisher Scientific
Incubator, 250 Cr Co2 Stack Adapter Plate, Cleanroom-compatible, required for stacking i250 CR models
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
INCUBATOR 250 CR CO2 STACK ADAPTER PLATE 76529-588 50162632 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty