Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Heracell VIOS 160i CO₂ Incubators, Thermo Scientific

Supplier: Thermo Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Heracell i
51033774 51033723 50162971 50162970 51033722 51033720 50162969 51033719 51033547 50162972 51033771 51033556 51033557 51033546
76529-594EA 24270 USD InStock
76529-594 76529-596 76459-190 76459-192 76459-170 76459-194 76459-172 76459-196 76459-186 76459-198 76459-188 76459-166 76459-200 76459-168
Heracell VIOS 160i CO₂ Incubators, Thermo Scientific
Incubators CO₂ Incubators
Designed to achieve your next breakthrough. Better solutions for optimal cell growth, VIOS CO₂ incubators provide the ideal in-vitro environment.

  • Pick the CO₂ incubator that's right for your lab's needs
  • Ideal for scale-up research and development
  • Strengthened stainless steel models for increased weight capacity
  • Optional reinforced shelves for large capacity, low media level culturing

Better solutions for optimal cell growth: Our CO₂ incubators provide the ideal in-vitro environment. In-chamber fan gently distributes clean, humidified air throughout the chamber ensuring homogeneous conditions. Fast recovery of all parameters (temperature, CO₂, and humidity) in 10 minutes or less, for stable culturing conditions and consistent results. In-chamber HEPA continuously filters the entire chamber air volume every 60 seconds. Superior ISO 5 clean room air quality is achieved within the culture chamber within 5 minutes. Directly heated water reservoir providing stable high relative humidity levels, preventing culture desiccation CO₂ and optional N₂/O₂ gases are pre-humidified before entering the chamber, providing a more constant, uniform environment. Auto-stop function disables fan operation during door openings to minimize air exchange.

Ultimate Contamination Control and Prevention. HEPA Filtration. In-chamber HEPA continuously filters the entire chamber air volume every 60 seconds. Superior ISO 5 clean room air quality is achieved within the culture chamber within 5 minutes.

Steri-Run, high temperature defection cycle. 12 hour 180 °C cycle assures uniform 6-log sterilization on all chamber surfaces. Eliminates biological contaminants including fungal mold, vegetative and spore forms of bacterial, including mycoplasma. Optional Copper interior.

In situ sensors and probes. Dual temperature probes for over temperature protection with operational back up if primary failure. The TC180 CO₂ sensor with humidity compensation allows for fast recovery from a routine door opening. The IR180Si incorporates silicon MEMS emitter as IR source with internal auto calibration for longer and more stable operation than a traditional IR sensor. Optional O₂ control with advanced Zirconium oxide sensors. Temperature resistant sensors do not need to be removed during sterilization cycle.

The Heracell VIOS CO₂ incubator delivers the performance reliability, ease of operation, and value required to support a range of culturing needs from basic research to the demanding, leading-edge applications, so you're ready for whatever comes next. Better solutions for optimal cell growth. Our CO₂ incubators with non-corrosive stainless-steel or 100% copper interior chambers provide the ideal in-vitro environment.

Certifications: CE and CSA compliant.

Ordering information: Accessories not included.

Order Now
 

ORDER

SPECIFICATIONS

SERVICES


Services: Thermo Scientific provides several service options such as Extended Warranty, Extended Warranty plus on-site Annual Preventative Maintenance, and Total Care. To learn more about these services please search "Heracell CO2 Incubator Technical Services" in the VWR search field.
In addition, VWRCATALYST™ can provide an array of services such as on-site IQ, OQ, PQ, Customized Protocol Generation, Equipment Calibration, Asset Management, and Certification Services (US only). Please call 888-793-2300 or email VWRCATALYST™@vwr.com for further details.

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty