Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Vikan® Tube Brush Heads for Flex Rod, Medium Bristles, Remco

Supplier: Remco
Vikan®
53683 53652 53684 53653 53655 53686 53654 53682 53656
76530-070EA 16.89 USD InStock
76530-070 76530-068 76530-058 76530-066 76530-056 76530-064 76530-054 76530-062 76530-060
Vikan® Tube Brush Heads for Flex Rod, Medium Bristles, Remco
Brushes
Medium bristle tube brush heads are great for cleaning pipes and other small areas.

  • Polyester bristles
  • Available in 0.8" and 1.6" Ø
  • Length: 7.9"

Brush heads are best suited for use with flexible handles (5350 and 53525).

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty