Would you like to log in again?
Vikan® Small Hand Scrapers, Remco
Supplier: Remco
Vikan®
696177 696166
76530-226EA 5.81 USD InStock
76530-226 76530-224
Vikan® Small Hand Scrapers, Remco
Scrapers
This color-coded small hand scraper is an excellent solution for all bench top scraping needs.
  • Polypropylene construction
  • Seamless design
  • Length: 7.8"
It has no seams or cracks, which helps prevent bacterial growth.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty