Would you like to log in again?
Vikan® Stainless Steel Scrapers, Remco
Supplier: Remco
Vikan®
69719 69721 69724 69723 697266 69715 69711 69717 69726 697277 697166 69716 69713 69718 69728 697177 69712 697288 69722 69729 69725 697188 69714 69727
76530-248EA 10.2 USD InStock
76530-248 76530-268 76530-246 76530-258 76530-266 76530-244 76530-288 76530-278 76530-256 76530-286 76530-264 76530-254 76530-276 76530-284 76530-262 76530-274 76530-252 76530-260 76530-282 76530-272 76530-250 76530-280 76530-270 76530-290
Vikan® Stainless Steel Scrapers, Remco
Scrapers
Color-coded stainless steel scrapers are an excellent solution for removing stubborn debris from smooth surfaces.
  • Stainless steel blade, polypropylene grip
  • Available in two widths: 1.5" and 3"
  • Length: 8.9"
The scraper’s beveled blade loosens dried pastry dough, starches, chocolate, and burned food. The scraper is made entirely of FDA compliant materials, making it safe for food contact. The ridged blade and ergonomic handle stand up to heavy and repeated use.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty