Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Vikan® UST Deck Scrub Brushes, Stiff Bristles, Remco

Supplier: Remco
Vikan®
70644 70648 70646 70641 70647 70645 70642 70643
76530-358EA 59.23 USD InStock
76530-358 76530-348 76530-356 76530-346 76530-354 76530-352 76530-350 76530-360
Vikan® UST Deck Scrub Brushes, Stiff Bristles, Remco
Brushes
UST 16" deck scrub brushes with stiff bristles.

  • Polyester bristles, polypropylene block
  • Euro (hygienic) thread type

FDA compliant.

Overall dimensions: 15.25L×1.88W×3.9"H

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty