Would you like to log in again?
Vortex Mixers, Crystal
Supplier: Crystal
VM-01UA VM-02UA
76531-200EA 304.65 USD InStock
76531-200 76531-202
Vortex Mixers, Crystal
Shakers and Mixers Vortex Mixers
Crystal Vortex Mixers are an ideal choice for reliable vigorous mixing, with simple touch start, continuous operation or timed modes and easily interchangeable platforms to accommodate many different tubes and plates.
  • Universal tube head for a wide variety of mixing applications
  • Three operational modes: Touch control, timed or continuous
  • Constant torque within full range of speed
  • Counter balance motor design for minimal vibration
  • Wide range of easy to change accessories to accommodate your sample container
  • 2 year replacement warranty
Versatile, with accessories to hold microplates, PCR tubes, combination of tube sizes and the option to customize your holder allows you to vortex many different sample containers with 1 instrument. Designed for years of trouble-free use.
Certifications: IP protection class according to DIN EN 60529: IP 21
CE marked
Delivery information: Supplied with single tube head (76531-204).
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty