Would you like to log in again?
MicroPolx® 4000-HP Ultrasonically Sealed Edge Wipes, Berkshire
Supplier: Berkshire
MP4000HP090910
76531-306CS 677.31 USD InStock
76531-306
MicroPolx® 4000-HP Ultrasonically Sealed Edge Wipes, Berkshire
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Designed for superior particle and bacterial removal without damaging critical surfaces.
  • Ultrasonically sealed edges minimize fiber and particle generation
  • High abrasion resistance
  • Soft hand and non-abrasive sealed-edge are excellent for sensitive or scratch prone surfaces
  • Microfiber yarn increases surface area and provides superior wiping efficiency and sorptive performance
MicroPolx® 4000-HP is a sealed edge cleanroom laundered wipe recommended for ISO Class 4 and above environments. The wipe is composed of polyester and nylon micro denier knitted fiber. It provides superior particle and bacterial removal without damaging critical surfaces. The high fiber surface area results in more efficient cleaning performance and minimizes the need for potentially harmful solvents.
Certifications: ISO Class 4 – 5. Cleanroom Class 10 – 100. EU Grade B/C/D.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty