Would you like to log in again?
Beakers, Low Form, Heavy Duty, United Scientific Supplies
Supplier: United Scientific Supplies
BG1003-600 BG1003-250 BG1003-1000 BG1003-50 BG1003-2000 BG1003-4000 BG1003-100 BG1003-400 BG1003-150
76531-386PK 83.65 USD InStock
76531-386 76531-388 76531-378 470349-762 76531-382 76531-384 76531-390 76531-380 76531-392
Beakers, Low Form, Heavy Duty, United Scientific Supplies
Beakers
Made from low-expansion borosilicate glass with extra-thick walls, these beakers are ideal for tough handling conditions, including mechanized washing.
  • Include white printed graduations (from 20 - 80 ml, at 10 ml intervals) and durable marking area
  • Tolerance is approximately +/- 5%
Available in multiple sizes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty