Would you like to log in again?
Burette, Class B, Glass Stopcock, United Scientific Supplies
Supplier: United Scientific Supplies
BR2124-25 BR2124-50 BR2124-10
76531-474EA 27.19 USD InStock
76531-474 76531-476 76531-472
Burette, Class B, Glass Stopcock, United Scientific Supplies
Burets
Made from high-quality accurate bore tubing for a high degree of accuracy, reliability and durability.
  • Amber graduations
  • Available in multiple sizes
Includes straight bore glass stopcock.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty