Would you like to log in again?
Crucibles, Glass, United Scientific Supplies
Supplier: United Scientific Supplies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CGQ050-L CGQ100-L CGQ080 CGQ100 CGQ015-L CGQ025-L CGQ150-L CGQ015 CGQ050 CGQ025 CGQ150 CGQ080-L
76531-522EA 35.6 USD InStock
76531-522 76531-534 76531-524 76531-536 76531-540 76531-530 76531-542 76531-520 76531-532 76531-526 76531-538 76531-528
Crucibles, Glass, United Scientific Supplies
Crucibles
Crafted from clear quartz glass, offering low expansion, dimensional stability and thermal shock resistance.
  • Maximum operating temperature: 1050 °C (1922 °F)
  • Quartz lid sold separately
Ideal for analytical applications that require ashing of samples.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty