Would you like to log in again?
Graduated Cylinders, Glass, Class A, Batch Certified, United Scientific Supplies
Supplier: United Scientific Supplies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CY3021-10 CY3021-1000 CY3021-2000 CY3021-25 CY3021-100 CY3021-50 CY3021-500 CY3021-250
76531-610EA 15.12 USD InStock
76531-610 76531-612 76531-602 76531-608 76531-614 76531-604 76531-616 76531-606
Graduated Cylinders, Glass, Class A, Batch Certified, United Scientific Supplies
Cylinders Measuring Cylinders
Made from borosilicate glass, with pour spout, heavy, uniform walls and stable hexagonal base. Bumper guard included.
  • Features double metric scale
Cylinder complies with ASTM E1272, Class A standards. Batch certified and calibrated to contain.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty