Would you like to log in again?
Disposable Culture Tubes, Plain, Borosilicate Glass, United Scientific Supplies
Supplier: United Scientific Supplies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
DCT051-1300 DCT051-1610 DCT051-1075 DCT051-1850 DCT051-25150 DCT051-1625 DCT051-1585 DCT051-1650 DCT051-1275 DCT051-0650 DCT051-20150
76531-632CS 60.99 USD InStock
76531-632 76531-644 76531-634 76531-646 76531-650 76531-640 76531-630 76531-642 76531-636 76531-648 76531-638
Disposable Culture Tubes, Plain, Borosilicate Glass, United Scientific Supplies
Tubes Culture Tubes
Disposable culture tubes manufactured from high quality 5.1 expansion borosilicate glass.
  • Rimless design for easy plugging with stopper (not included)
  • Packaged in shrinkwrapped boxes
Caps not included.
Certifications: Meet USP Type I and ASTM E438, Type I, Class B requirements.
Packaging: Tubes are packaged in shrink-wrapped boxes to keep them particle-free, and four boxes are included in each case.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty