Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Sunday, May 25. 
Boiling Flask, Flat Bottom, Ground Joints, United Scientific
Supplier: United Scientific Supplies
FFB058-3000 FFB058-500 FFB058-5000 FFB058-2000 FFB058-250 FFB058-50 FFB058-100 FFB058-1000
76531-666PK 147.9 USD InStock
76531-666 470339-920 470339-558 76531-668 470339-756 76531-672 76531-670 470339-812
Boiling Flask, Flat Bottom, Ground Joints, United Scientific
Flasks Flat Bottom Flasks
Boiling flasks with ground glass joints are made from low-expansion borosilicate glass.
  • Uniform wall thickness
  • Ground glass joints 24/40
Boiling flasks with ground glass joints feature uniform wall thickness for superior mechanical strength and shock resistance. All vessels have a joint size of 24/40.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty