Would you like to log in again?
Petri Dishes, Borosilicate Glass, United Scientific Supplies
Supplier: United Scientific Supplies
G1075 G1100 G1060
76532-014PK 74.94 USD InStock
76532-014 76532-012 76532-016
Petri Dishes, Borosilicate Glass, United Scientific Supplies
Culture Dishes Petri Dishes
Pack of 10 autoclavable borosilicate glass petri dishes.
  • Crafted from high-quality borosilicate glass
  • Autoclavable
Lids are included. Note: lids do not seal air-tight.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty