Would you like to log in again?
Pipettes, Measuring (Mohr), Class A, Batch Certified, United Scientific Supplies
Supplier: United Scientific Supplies
PM7059-H PM7059-J PM7059-A PM7059-B PM7059-D PM7059-I PM7059-F
76532-068EA 1.2 USD InStock
76532-068 76532-058 76532-066 76532-060 76532-070 76532-064 76532-062
Pipettes, Measuring (Mohr), Class A, Batch Certified, United Scientific Supplies
Pipettes Measuring (Mohr) Pipettes
Color-coded Mohr pipettes.
  • Printed with durable amber markings
  • Color-coded in white for easy identification of capacity
Made from borosilicate glass in compliance with ASTM specification E1293. Batch certified and calibrated to deliver.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty