Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Pipettes, Measuring (Mohr), Class B, United Scientific Supplies
Supplier: United Scientific Supplies
PM7060-A PM7060-D PM7060-B PM7060-F PM7060-C PM7060-H PM7060-I
76532-078PK 20.33 USD InStock
76532-078 76532-082 76532-072 76532-080 76532-076 76532-084 76532-074
Pipettes, Measuring (Mohr), Class B, United Scientific Supplies
Pipettes Measuring (Mohr) Pipettes
Color-coded Mohr pipettes.
  • Printed with durable amber markings
  • Color-coded in white for easy identification of capacity
Made from borosilicate glass in compliance with ASTM specification E1293 for Class B pipettes. Calibrated to deliver.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty